Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminohippuric acid for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.14621.0025
EM8.14621.0025CS 20928 USD InStock
EM8.14621.0025
4-Aminohippuric acid for synthesis, Sigma-Aldrich®
4-Aminohippuric acid


Formula: H₂NC₆H₄C(O)NHCH₂COOH
MW: 194.19 g/mol
Boiling Pt: 441
MDL Number: MFCD00007890
CAS Number: 61-78-9
Merck Index: 13,00441
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 199 - 202 °C (decomposition)
Molar Mass 194.18 g/mol
Bulk Density 380 kg/m3
pH 3.0 - 3.5 (20 g/l, H2O, 20 °C)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty