Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.18233.0001
EM8.18233.0001CS 146831 USD InStock
EM8.18233.0001
(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one for synthesis, Sigma-Aldrich®
(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one


Formula: C₂₁H₁₈O₇
MW: 382.368 g/mol
CAS Number: 25552-06-1
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 124 - 127 °C
Molar Mass 382.37 g/mol
Synonyms Methyl-3,6-dibenzoyl-2-deoxy-α-D-glycero-hex-2-enopyranosid-4-ulose
Hill Formula C21H18O7

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty