Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.18378.0025
EM8.18378.0025CS 11617 USD InStock
EM8.18378.0025
(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®
(S)-(-)-β-Citronellol


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
MDL Number: MFCD00063214
CAS Number: 7540-51-4
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Molar Mass 156.27 g/mol
Boiling Point 109 - 111 °C (13 hPa)
Flash Point 101 °C DIN 51758
Refractive Index 1.4562 (20 °C, 589 nm)
Density 0.855 g/cm3 (20 °C)
Ignition Point 240 °C
Synonyms (S)-(-)-3,7-Dimethyl-6-octen-1-ol, ß-Rhodinol
Hill Formula C10H20O

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty