Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethyl-3-hexyne-2,5-diol for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.18587.0100
EM8.18587.0100CS 1134 USD InStock
EM8.18587.0100
2,5-Dimethyl-3-hexyne-2,5-diol for synthesis, Sigma-Aldrich®
2,5-Dimethyl-3-hexyne-2,5-diol


Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 205
MDL Number: MFCD00004468
CAS Number: 142-30-3
Order Now
Specification Test Results
Solubility 240 g/l (20 °C)
Melting Point 93 - 95 °C
Molar Mass 142.19 g/mol
Boiling Point 205 °C (1013 hPa)
Density 0.94 g/cm3 (20 °C)
Hill Formula C8H14O2

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty