Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropylsilane for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

MQ200

8.41359.0010
EM8.41359.0010CS 77663 USD InStock
EM8.41359.0010
Triisopropylsilane for synthesis, Sigma-Aldrich®
Triisopropylsilane


Formula: C₉H₂₂Si
MW: 158.36 g/mol
Boiling Pt: 84
Density: 0.7734 g/cm³ (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00009657
CAS Number: 6485-79-6
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Molar Mass 158.35 g/mol
Boiling Point 84 - 85 °C (46.7 hPa)
Flash Point 35 °C
Density 0.77 g/cm3 (20 °C)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty