Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyvinylpyrrolidone, M.W. 40000 avg, white powder

Supplier: E. Becker & Co. GmbH

0219545180 0219545190 0219545191
IC19545180EA 77.9 USD InStock
IC19545180 IC19545190 IC19545191
Polyvinylpyrrolidone, M.W. 40000 avg, white powder
Polyvinylpyrrolidone
Polyvinylpyrrolidone is a water-soluble polymer made from the monomer N-vinylpyrrolidone. Typically used as thickeners, dispersing agents, detoxicant, complexing agent, lubricants and binders.

Polyvinylpyrrolidone is used as an emulsifier and disintegrant for solution polymerisation; also for production of membranes, such as dialysis and water purification filters; as an aid for increasing the solubility of drugs in liquid and semi-liquid dosage forms (syrups, soft gelatine capsules) and as an inhibitor of recrystallisation.

Complexes with phenolics and alkaloids for their removal from plant samples, thus preventing their modification of proteins and any interference they may cause in spectrophotometric determinations of protein content. This is also reported to improve stability of enzymes.

Solubility ( 2,5% aq soln): clearly soluble, free of floating particles.

Polyvinylpyrrolidone is a water-soluble polymer made from the monomer N-vinylpyrrolidone. Typically used as thickeners, dispersing agents, detoxicant, complexing agent, lubricants and binders.

Precaución: Research Use Only (RUO).


Formula: (C₆H₉NO)n
Boiling Pt: 90…93 °C
Melting Pt: 13.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c43b4a0acc619e9792a26d5b560f0211ac60b9&vid=b0c43b4a0acc619e9792a26d5b560f0211ac60b9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
FTIR Conforms to Standard
pH (5% aq soln) 2.8 - 4.2
Solubility ( 2.5% aq soln) Clearly Soluble, Free of Floating Particles

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty