Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, white powder for electrophoresis

Supplier: E. Becker & Co. GmbH
Warning

04856126
IC856126EA 224.48 USD InStock
IC856126
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, white powder for electrophoresis
Dithiothreitol (DTT, Cleland's reagent)
DL-Dithiothreitol is also known as Clelands reagent; Protective agent for sulfhydryl groups (-SH). Quantitatively reduces disulfides (-S-S- to -SH). In this reaction the DTT is oxidized to the cyclic disulfide which ensures the reduction of other disulfides in solution. Disulfide reduction occurs quickly at pH 8.
Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes. Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol.
Useful for stabilizing sulfhydryl-containing enzymes.

  • Electrophoresis Grade
  • Half Life: 1.4( pH 8.5 at 20 °C)
  • pH: 4.0 to 6.0 (0.1 M in H2O, 25 °C)
  • Redox potential = -0.33 volts at pH 7
  • Soluble in water (50 mg/mL - clear and colorless solution); soluble in ethanol, acetone, ethylate, chloroform and ether.
  • +4 °C

Sulfhydryl reducing agent Blotting Protein Purification

DL-Dithiothreitol is also known as Clelands reagent; Protective agent for sulfhydryl groups (-SH). Quantitatively reduces disulfides (-S-S- to -SH). In this reaction the DTT is oxidized to the cyclic disulfide which ensures the reduction of other disulfides in solution. Disulfide reduction occurs quickly at pH 8. Half life at pH 6.5 and 20 °C was found to be 40 hours.

Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes. Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).It is used in preparation of carboxymethylated rPA


Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Melting Pt: 40…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
UN: 3335
ADR: 9 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=0c72cc68bbc36ab17baa927731844d74085edbf9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99.5%
FTIR Conforms to Standard

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty