Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Hexane 95 GC, corrected for water content HPLC/UHPLC, Spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger

M.O.C. Category NI

9304-02 9304-33 9304-03 9304-50 9304-63 9304-7Y 9304-99 9304-H1 9304-ZV
JT9304-2CS 1029.15 USD InStock
JT9304-2 JT9304-33 JT9304-3 JT9304-50 JT9304-63 JT9304-7Y JT9304-88 JT9304-99 JT9304-H1 JT9304-ZV
n-Hexane 95 GC, corrected for water content HPLC/UHPLC, Spectrophotometry, J.T.Baker®
n-Hexane
For use in liquid chromatography and spectrophotometry. UV cutoff 192 nm. Lot analysis on label.


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c939cebcd37af6287105861069a46c7635aa4af8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.008
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.2
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤192
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥99.5 %
Assay (as n-Hexane) (by GC, corrected for water) ≥95.0 %
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.01 %
Filtered through a 0.2 micron filter.
Packaged under Nitrogen.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty