Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetic acid glacial ≥99.7%, CMOS for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger

Management of Change (MOC) category = R

9513-07
JT9513-7EA 409.7 USD InStock
JT9513-7 JT9513-9
Acetic acid glacial ≥99.7%, CMOS for microelectronic, J.T.Baker®
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=031896546f0eb78526a541f0af4636e3991e9f8e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Microelectronic Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
Assay (CH₃COOH) (by freezing point) ≥99.7 %
Color (APHA) ≤10
Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
Acetaldehyde ≤0.05 %
Residue after Evaporation ≤4 ppm
Solubility in H₂O Passes Test
Specific Gravity at 20°/20°C ≥1.048
Substances Reducing Dichromate Passes Test
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤1 ppm
Phosphate (PO₄) ≤1 ppm
Sulfate (SO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤50 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤30.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤20.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤10.0 ppb
Heavy Metals (as Pb) ≤300.0 ppb
Trace Impurities - Iron (Fe) ≤100.0 ppb
Trace Impurities - Lead (Pb) ≤100 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤25.0 ppb
Trace Impurities - Niobium (Nb) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤100 ppb
Trace Impurities - Silicon (Si) ≤50 ppb
Trace Impurities - Silver (Ag) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤100.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤50.0 ppb
Trace Impurities - Tin (Sn) ≤50 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤50 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty