Cadmium Reduction Column, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
419000-8505
KT419000-8505EA 372.84 USD InStock
KT419000-8505
Cadmium Reduction Column, Kimble Chase
Chromatography Columns
This column features all borosilicate glass construction with an 85 ml reservoir.
Ref: Method 4500-NO3-E of the Standards Methods for the Examination of Water and Wastewater. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The glass wool plug and PTFE tubing at the bottom of the column are held securely in place by a BEVEL-SEAL™ connection, facilitating easy cleaning and assembly. PTFE BEVEL-SEAL™ stopcock connector is included as a metering valve to control flow rate. Stopcock has a plug (glassMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty