Extractor Body, Kimble-chase
Supplier: DWK Life Sciences (KIMBLE)
584191-0000
KT584191-0000EA 650.13 USD InStock
KT584191-0000
Extractor Body, Kimble-chase
Extractors Liquid-Liquid Extractors
The extractor body is designed for EPA priority pollutant samples where the extracting solvent is heavier than water.
  • Extractor body has a standard taper 45/50 top joint and a 24/40 side joint
  • Outer Ø is about 100 mm
Mainly designed for samples of one liter plus approximately 200 ml of extracting solvent.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty