KIMBLE® Distilling Flasks, Round Bottom, 5-Neck, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
607500-1024
KT607500-1024EA 369.92 USD InStock
KT607500-1024
KIMBLE® Distilling Flasks, Round Bottom, 5-Neck, DWK Life Sciences
Flasks Distillation Flasks
Five-neck heavy wall round-bottom flask with vertical standard taper outer joints.
  • Side necks are 90° apart
  • Where possible, vertical necks will be supplied
  • Manufactured from 33 expansion, low extractable borosilicate glass
On small flasks having large necks, positioning may have to be made at an angle.
Certificaciones: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty