Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Supplier: VWR International
MFLX75219-00 MFLX75219-20 MFLX75219-40 MFLX75219-50 MFLX75219-60 MFLX75219-05 MFLX75219-10 MFLX75219-15 MFLX75219-55
MFLX75219-00EA 3450 USD InStock
MFLX75219-00 MFLX75219-20 MFLX75219-40 MFLX75219-50 MFLX75219-60 MFLX75219-05 MFLX75219-10 MFLX75219-15 MFLX75219-55
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Pumps Drum/Transfer Pumps
Capable of pumping fluids with viscosities up to 1000 cps.
  • 316 SS construction with PTFE elastomers
  • Easy to clean with Tri-Clamp® fittings
The sanitary drum pumps are engineered to transfer low-viscosity food products, pharmaceutical materials, and personal care solutions from drums, totes, and kettles. Wetted surfaces are constructed with FDA-compliant materials which provide an ideal solution for sanitary applications.
Información de suministro: Includes drive and pump tube.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty