Masterflex® I/P® Brushless Process Drive with Remote Input/Output, Avantor®
Supplier: MASTERFLEX LLC
77411-00 77411-00 77411-00 77411-00 77411-00 77411-00 77411-00 77411-00 77411-00
MFLX77411-00EA 5190 USD InStock
MFLX77411-00 MFLX77411-00-AU MFLX77411-00-DK MFLX77411-00-IN MFLX77411-00-IL MFLX77411-00-EU MFLX77411-00-UK MFLX77411-00-CH MFLX77411-00-IT
Masterflex® I/P® Brushless Process Drive with Remote Input/Output, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Powerful, low-maintenance drives are versatile and cost-effective.
  • Flow rate 0.2 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Contructed of non-chip painted steel and aluminum IP55-rated housing
  • Brushless, maintenance-free motor offers ±0.25% speed control accuracy
  • Remote control available using analog
More Product Information
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty