µ-Dish 35 mm, High
Supplier: IBIDI USA MS
81156
MSPP-81156PK 400 USD InStock
MSPP-81156
µ-Dish 35 mm, High
Culture Dishes Cell Culture Dishes
A 35 mm imaging dish with a polymer coverslip bottom for high-end microscopy and cell-based assays.
  • Equipped with a lid with locking feature that minimizes evaporation
  • Compaitable with a DIC Lid
  • Biocompatible plastic material—no glue, no leaking
  • Observation area of 21 mm
In this microscopy dish, the cells are imaged on a No. 1.5 ibidi Polymer Coverslip bottom with the highest optical quality and optimal cell adherence. This is a cell culture dish for everyday use with diverse application possibilities.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty