Air Science USA® Airflow Meter for Ductless Fume Hoods, Air Science USA LLC
Supplier: Air Science
DWYER
MSPP-DWYEREA 83.08 USD InStock
MSPP-DWYER
Air Science USA® Airflow Meter for Ductless Fume Hoods, Air Science USA LLC
Fume Hoods and Enclosures Ductless Fume Hoods
Side wall mounted air flow indicator for ductless fume hoods.
  • Flexible mounting
  • Easy to read
  • Never needs to be calibrated
Velometer, continuous display of face velocity, side wall mounted.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty