Pierce™ Inclusion Body Solubilization Reagent, Thermo Scientific
Supplier: Paul Schöndorf Metallwaren GmbH
78115
PI78115EA 317.48 USD InStock
PI78115
Pierce™ Inclusion Body Solubilization Reagent, Thermo Scientific
Protein Purification Protein Purification Reagents
Inclusion Body Solubilization Reagent effectively retrieves recombinant proteins that form insoluble aggregates during bacterial overexpression.
  • Achieves the essential first step for subsequent insoluble protein refolding procedures
  • One easy to use solution
  • Compatible with Coomassie Plus Protein Assay Kit to determine degree of expression
  • Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE can be performed
  • Can accommodate additional components such as reducing agents or chelating agents
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty