Pierce™ High pH Reversed-Phase Peptide Fractionation Kit, Thermo Scientific
Supplier: Paul Schöndorf Metallwaren GmbH
84868
PI84868EA 438.15 USD InStock
PI84868
Pierce™ High pH Reversed-Phase Peptide Fractionation Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
Thermo Scientific™ Pierce™ High pH Reversed-Phase Peptide Fractionation Kit increases protein identification from LC/MS analysis through orthogonal peptide fractionation of complex peptide samples.Easy-to-use—resin provided in single-use spin column formatImproved protein identifications—protein identifications increased by ≥50% when compared to unfractionated samplesReproducible—elution profiles and fractional resolution vary by less than 20%Optimized—robust procedure for maximal protein identification and peptide recovery while minimizing fractional overlapCompatible—reagents validated with a variety of complex samples, including TMT™-labeled peptidesIn order to enable deep proteome sequencing, it is often necessary to reduce the sample complexity by fractionation in an orthogonal dimension prior to LC/MS analysis. The Pierce High pH Reversed-Phase Peptide Fractionation Kit utilizes high pH reversed-phase chromatography to separate peptides by hydrophobicity and provides excellent orthogonality to low pH reversed-phase LC-MS gradients.The kit has been designed to improve protein identification through the use of a proprietary reversed-phase resin in an easy-to-use spin column format with a highpH fractionation protocol. In contrast to strong cation exchange (SCX) fractionation, the high-pH reversed-phase fractions do not require an additional desalting step before LC/MS analysis.The High pH Reversed-phase Peptide Fractionation Kit includes a high pH buffer (0.1% triethylamine) and twelve spin columns containing pH-resistant reversed-phase resin. Each reversed-phase fractionation spin column enables fractionation of 10–100 µg of peptide sample using a microcentrifuge.Native, phosphorylated, Tandem Mass TagTM (TMTTM)-labeled, and other complex peptide mixture samples can be fractionated using this kit. Combining the search results generated by the individual fractions helps improve protein sequence coverage and increases the number of identified proteins relative to unfractionated samples.Applications:Reducing sample complexity to identify targets of interestConducting systems biology studiesReducing sample complexity to improve quantitation studies
  • Easy-to-use - resin provided in single-use spin column format
  • Improved protein identifications - protein identifications increased by ≥50% when compared to unfractionated samples
  • Reproducible - elution profiles and fractional resolution vary by less than 20%
  • Optimized - robust procedure for maximal protein identification and peptide recovery while minimizing fractional overlap
  • Compatible - reagents validated with a variety of complex samples, including TMT™-labeled peptides
Información para pedidos: For Research Use Only. Not for use in diagnostic procedures. Reversed-phase fractionation spin columns, 12 columns; Triethylamine (0.1%), 100 mL; Store in refrigerator (2–8˚C).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty