Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mercury(II) nitrate 0.0250 Normal (N/40) titrant

Supplier: RICCA CHEMICAL CO
Danger

R4715000-1C
RC4715-32EA 122 USD InStock
RC4715-32
Mercury(II) nitrate 0.0250 Normal (N/40) titrant
Mercury(II) nitrate
For EPA and ASTM testing.


Formula: HgN₂O₆
MW: 324.6
Melting Pt: -38.9
Density: 4.3 g/cm³ (20 °C)
MDL Number: MFCD00011038
CAS Number: 10045-94-0
EINECS: 233-152-3
UN: 1625
ADR: 6.1 II
Merck Index: 12,05933
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.02500 ± 0.00001 N at 20.0 °C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty