Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Water ASTM Type IV

Supplier: RICCA CHEMICAL CO

R9151400-1A R9151400-20F R9151400-55E
RCR9151400-1AEA 82 USD InStock
RCR9151400-1A RCR9151400-20F RCR9151400-55E
Water ASTM Type IV
Water


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098
Order Now
Specification Test Results
Appearance Colorless liquid
Conductivity at 25°C max 5.0 µS/cm
pH at 25°C 5.0-8.0
Sodium (Na) max 50 ppb
Chloride (Cl) max 50 ppb

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty