VeriSpec® Calcium Standard for ICP/MS 100 ppm in 2% HNO₃
Supplier: RICCA CHEMICAL CO
RV010510-100N
RCRV010510100NEA 564 USD InStock
RCRV010510100N
VeriSpec® Calcium Standard for ICP/MS 100 ppm in 2% HNO₃
Standards Cationic Standards Calcium Standards
Certified reference material (CRM) for ICP-MS
Certificaciones: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty