TES buffer, 1X, ready to use, pH 7.8
Supplier: Bandelin Electronic GmbH &Co.KG
MB-058
RLMB-058EA 145.22 USD InStock
RLMB-058
TES buffer, 1X, ready to use, pH 7.8
Buffers Biological Buffers
This product is ready-to-use as a working 1X solution and requires no further dilution
This TES buffer consists of 10 mM Tris HCl, 100 mM NaCl, and 1.0 mM EDTA at a pH of 7.8. Meticulously prepared using ultra pure reagents dissolved in DEPC treated pharmaceutical grade deionized water.
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Precaución: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty