Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-3-Octene ≥98.0%

Supplier: Decon Laboratories Ltd

O0133-5ML
TCO0133-005MLEA 625 USD InStock
TCO0133-005ML
cis-3-Octene ≥98.0%
cis-3-Octene
CAS Number: 14850-22-7
MDL Number: MFCD00067486
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 17
Specific Gravity (20/20): 0.72


Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 123
Melting Pt: -126
Density: 0.72 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00067486
CAS Number: 14850-22-7
EINECS: 238-912-8
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty