Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc trifluoromethanesulphonate ≥98.0%

Supplier: Decon Laboratories Ltd

T1294-25G
TCT1294-25GEA 75.02 USD InStock
TCT1294-25G
Zinc trifluoromethanesulphonate ≥98.0%
Zinc trifluoromethanesulphonate
CAS Number: 54010-75-2
MDL Number: MFCD00013229
Molecular Formula: CHF3O3S
Molecular Weight: 363.51
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Stable Lewis Acids in Aqueous Media
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid Zinc(II) Salt, Zinc(II) Triflate


Formula: C₂F₆O₆S₂Zn
MW: 363.53
MDL Number: MFCD00013229
CAS Number: 54010-75-2
UN: 3261
ADR: 8 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afbacc3c722a7c7ae47fb21f186cddd978646ee5&vid=afbacc3c722a7c7ae47fb21f186cddd978646ee5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty