Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20901-0500
TS20901-0500EA 128 USD InStock
TS20901-0500
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate
Propanephosphonic acid cyclic anhydride


Formula: C₉H₂₁O₆P₃
MW: 318.19 g/mol
Boiling Pt: 77.2 °C
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006583
CAS Number: 68957-94-8
EINECS: 422-210-5
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to amber Solution
Assay ≥50 % w/w Solids

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty