Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetone-[D₆] 99.8 atom % D, AcroSeal™ for NMR spectroscopy

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21737-1000
TS21737-1000EA 455 USD InStock
TS21737-1000
Acetone-[D₆] 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Acetone-[D₆]


Formula: D₃CC(O)CD₃
MW: 64.03 g/mol
Boiling Pt: 55.5
Melting Pt: -95.4
Density: 0.87 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
UN: 1090
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.75 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty