Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

396890010 396891000
TS396890010EA 187 USD InStock
TS396890010 TS396891000
Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal™
Ethylene glycol monomethyl ether (2-methoxyethanol)


Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124.6
Melting Pt: -85
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
UN: 1188
ADR: 3 III
Merck Index: 12,06118
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.003 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty