Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(tert-Butyldimethylsilyloxy)phenylboronic acid 96+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39834-0010
TS39834-0010EA 89 USD InStock
TS39834-0010 TS39834-0050
3-(tert-Butyldimethylsilyloxy)phenylboronic acid 96+%
3-(tert-Butyldimethylsilyloxy)phenylboronic acid


Formula: C₁₂H₂₁BO₃Si
MW: 252.19 g/mol
Melting Pt: 86…91 °C
Storage Temperature: Ambient
MDL Number: MFCD03701505
CAS Number: 261621-12-9
Order Now
Specification Test Results
Appearance Off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty