VWR® Square Media Biocanisters
Supplier: VWR International
VWRI210-000105 VWRI210-000106 VWRI210-000107 VWRI210-000108 VWRI210-000109 VWRI210-000110
VWRI210-000105CS 268.48 USD InStock
VWRI210-000105 VWRI210-000106 VWRI210-000107 VWRI210-000108 VWRI210-000109 VWRI210-000110
VWR® Square Media Biocanisters
Bottles Plastic Bottles
VWR® square containers are ideal for storage of media, laboratory reagents, and biological buffers. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic containers.
  • Made from durable 100% virgin material
  • Leakproof bottles and HDPE closures
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability
All containers are graduated, and feature a molded hand grip for improved ergonomics.
Polycarbonate containers are suitable for use inMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty