Macro-Prep HIC Media, Bio-Rad
1580090 1560090 1580080 1560080
101643-088EA 246.15 USD InStock
101643-088 101643-034 101643-086 101643-030
Macro-Prep HIC Media, Bio-Rad
Protein Purification Protein Purification Resins
Macro-Prep hydrophobic interaction chromatography (HIC) media are methacrylate-based 50µm beads for protein, polypeptide, enzyme, and nucleic acid purification
These media are available in two functional forms: the weakly hydrophobic methyl support is ideal for purifying compounds that have strong hydrophobic regions and the mildly hydrophobic t-butyl support is ideal for purifying compounds that have few or weakly hydrophobic regions. Both media are chemically and thermally stable. They are autoclavable and can withstand treatment in acid, base, chaotropic agents, or detergents while retaining high protein binding capacities. They can be cleaned in place with ethanol or sodium hydroxide.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty