Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pentenyl acetate ≥98.0%

Supplier: Decon Laboratories Ltd

A1345-500ML A1345-25ML
TCA1345-500MLEA 587 USD InStock
TCA1345-500ML TCA1345-025ML
4-Pentenyl acetate ≥98.0%
4-Pentenyl acetate
CAS Number: 1576-85-8
MDL Number: MFCD00008723
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 45
Specific Gravity (20/20): 0.91

Other Product Names / Synonyms: Acetic Acid 4-Pentenyl Ester, 5-Acetoxy-1-pentene


Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 151 °C
Density: 0.91 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00008723
CAS Number: 1576-85-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=476ec96a2f7474e00978cccee00c7ede72f04f15&vid=476ec96a2f7474e00978cccee00c7ede72f04f15&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty