Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

B1631-5G B1631-25G
TCB1631-005GEA 43 USD InStock
TCB1631-005G TCB1631-025G
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine
CAS Number: 2130-96-3
MDL Number: MFCD00065597
Molecular Formula: C21H25NO5
Molecular Weight: 371.43
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 113
Specific rotation [a]20/D: 29.5 deg (C=2, EtOH)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-O-benzyl-L-tyrosine, Boc-Tyr(Bzl)-OH


Formula: C₂₁H₂₅NO₅
Melting Pt: 113 °C
MDL Number: MFCD00065597
CAS Number: 2130-96-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab9895c5901945c0db770ab218059fc08ea5fdc5&vid=ab9895c5901945c0db770ab218059fc08ea5fdc5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty