Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-nitropyridine ≥98.0%

Supplier: Decon Laboratories Ltd

C1765-5G C1765-1G
TCC1765-5GEA 157 USD InStock
TCC1765-5G TCC1765-1G
3-Chloro-2-nitropyridine ≥98.0%
3-Chloro-2-nitropyridine
CAS Number: 54231-32-2
MDL Number: MFCD00490110
Molecular Formula: C5H3ClN2O2
Molecular Weight: 158.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91


Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00490110
CAS Number: 54231-32-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e19a4b025422538255496b1cff25889ef632884&vid=0e19a4b025422538255496b1cff25889ef632884&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty