Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-hydroxynicotinic acid ≥98.0%

Supplier: Decon Laboratories Ltd

C2339-1G C2339-5G
TCC2339-1GEA 26 USD InStock
TCC2339-1G TCC2339-5G
5-Chloro-2-hydroxynicotinic acid ≥98.0%
5-Chloro-2-hydroxynicotinic acid
CAS Number: 38076-80-1
MDL Number: MFCD00204158
Molecular Formula: C6H4ClNO3
Molecular Weight: 173.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 262

Other Product Names / Synonyms: 5-Chloro-2-hydroxy-3-pyridinecarboxylic Acid


Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
Melting Pt: 262 °C
MDL Number: MFCD00204158
CAS Number: 38076-80-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c9612f67a13d93fc73a6732af31fdd599fa7b5b&vid=1c9612f67a13d93fc73a6732af31fdd599fa7b5b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty