Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Xylylenediamine ≥99.0%

Supplier: Decon Laboratories Ltd

D1018-500G D1018-25G
TCD1018-500GEA 936 USD InStock
TCD1018-500G TCD1018-25G
p-Xylylenediamine ≥99.0%
p-Xylylenediamine
CAS Number: 539-48-0
MDL Number: MFCD00009821
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 62
Flash Point (°C): 142

Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: alpha,alpha'-Diamino-p-xylene, p-Xylene Diamine, 4-(Aminomethyl)benzylamine


Formula: C₈H₁₂N₂
MW: 136.20 g/mol
Boiling Pt: 125 °C
Melting Pt: 62 °C
Flash Pt: 142 °C
MDL Number: MFCD00009821
CAS Number: 539-48-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56ecd4e715b5335d16b4506bda5e590cab798c11&vid=56ecd4e715b5335d16b4506bda5e590cab798c11&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty