Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Decyltrimethylammonium bromide ≥99.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D1467-25G D1467-500G
TCD1467-025GEA 55.97 USD InStock
TCD1467-025G TCD1467-500G
Decyltrimethylammonium bromide ≥99.0% (by titrimetric analysis)
Decyltrimethylammonium bromide
CAS Number: 2082-84-0
MDL Number: MFCD00041973
Molecular Formula: C13H30BrN
Molecular Weight: 280.29
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 242


Formula: C₁₃H₃₀BrN
Melting Pt: 242 °C
MDL Number: MFCD00041973
CAS Number: 2082-84-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69ae61ae22a76633b7b5a98bc82661c963e595e6&vid=69ae61ae22a76633b7b5a98bc82661c963e595e6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty