Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-4-hydroxybenzonitrile ≥95.0%

Supplier: Decon Laboratories Ltd

D2790-1G D2790-5G
TCD2790-1GEA 19 USD InStock
TCD2790-1G TCD2790-5G
3,5-Dimethyl-4-hydroxybenzonitrile ≥95.0%
3,5-Dimethyl-4-hydroxybenzonitrile
CAS Number: 4198-90-7
MDL Number: MFCD00221716
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 125

Other Product Names / Synonyms: 4-Cyano-2,6-dimethylphenol, 2,6-Dimethyl-4-cyanophenol, 3,5-Dimethyl-4-hydroxybenzonitrile


Formula: C₉H₉NO
Melting Pt: 125 °C
MDL Number: MFCD00221716
CAS Number: 4198-90-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad7a3fd8bc67fdc6160cba0bfe5391f1198f1cc&vid=7ad7a3fd8bc67fdc6160cba0bfe5391f1198f1cc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty