Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl dodecanedioate ≥98.0%

Supplier: Decon Laboratories Ltd

D2835-25G D2835-500G
TCD2835-25GEA 53 USD InStock
TCD2835-25G TCD2835-500G
Dimethyl dodecanedioate ≥98.0%
Dimethyl dodecanedioate
CAS Number: 1731-79-9
MDL Number: MFCD00043632
Molecular Formula: C14H26O4
Molecular Weight: 258.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Other Product Names / Synonyms: Dodecanedioic Acid Dimethyl Ester


Formula: C₁₄H₂₆O₄
MDL Number: MFCD00043632
CAS Number: 1731-79-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d097bf4707cbdeb4121a071de7b7f782b777083&vid=6d097bf4707cbdeb4121a071de7b7f782b777083&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty