Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-(morpholino)propionate ≥97.0%

Supplier: Decon Laboratories Ltd

E0784-1G E0784-5G
TCE0784-1GEA 24 USD InStock
TCE0784-1G TCE0784-5G
Ethyl-3-(morpholino)propionate ≥97.0%
Ethyl-3-(morpholino)propionate
CAS Number: 20120-24-5
MDL Number: MFCD00014622
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 118
Specific Gravity (20/20): 1.04

Other Product Names / Synonyms: 3-(Morpholino)propionic Acid Ethyl Ester, 4-[2-(Ethoxycarbonyl)ethyl]morpholine


Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Boiling Pt: 118 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00014622
CAS Number: 20120-24-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e058913f8c29a2688091c165711f03bde3ee8e1b&vid=e058913f8c29a2688091c165711f03bde3ee8e1b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty