Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoropelargonic acid ≥95.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

H0843-25G H0843-5G
TCH0843-25GEA 236 USD InStock
TCH0843-25G TCH0843-5G
Perfluoropelargonic acid ≥95.0% (by GC, titration analysis)
Perfluoropelargonic acid
CAS Number: 375-95-1
MDL Number: MFCD00039605
Molecular Formula: C9HF17O2
Molecular Weight: 464.08
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 65

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Heptadecafluoropelargonic Acid, Perfluorononanoic Acid, Perfluoropelargonic Acid


Formula: C₉HF₁₇O₂
Melting Pt: 65 °C
MDL Number: MFCD00039605
CAS Number: 375-95-1
UN: 3261
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=defa4d463ed02ff1e8e159529801970e7f937766&vid=defa4d463ed02ff1e8e159529801970e7f937766&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty