N0091-500G
N0091-25G
TCN0091-500GEA
658
USD
InStock
TCN0091-500G
TCN0091-25G
Nicotinoyl chloride hydrochloride ≥95.0%
Nicotinoyl chloride hydrochloride
CAS Number: 20260-53-1
MDL Number: MFCD00012820
Molecular Formula: C6H4ClNO
Molecular Weight: 178.01
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 155
Other Product Names / Synonyms: Pyridine-3-carbonyl Chloride Hydrochloride