N0350-500G
N0350-25G
TCN0350-500GEA
259.4
USD
InStock
TCN0350-500G
TCN0350-025G
Nonenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0%
Nonenylsuccinic anhydride (mixture of branched chain isomers)
CAS Number: 28928-97-4
MDL Number: MFCD00014552
Molecular Formula: C13H20O3
Molecular Weight: 224.30
Purity/Analysis Method: 90.0% (T)
Form: Clear Liquid
Flash Point (°C): 147
Specific Gravity (20/20): 1.03
Form: Clear Liquid
Flash Point (°C): 147
Specific Gravity (20/20): 1.03