Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-1-Naphthylethylenediamine dihydrochloride monomethanolate ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

N0577-25G N0577-1G
TCN0577-025GEA 102 USD InStock
TCN0577-025G TCN0577-001G
N-1-Naphthylethylenediamine dihydrochloride monomethanolate ≥97.0% (by titrimetric analysis)
N-1-Naphthylethylenediamine dihydrochloride monomethanolate
MDL Number: MFCD00012556
Molecular Formula: C12H14N2
Molecular Weight: 291.22
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 190

Other Product Names / Synonyms: Bratton-Marshall Reagent, Marshall's Reagent


https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2beff6d975283f3c6446d4658fc27c9c7b4ba972&vid=2beff6d975283f3c6446d4658fc27c9c7b4ba972&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty