Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrochalcone ≥95.0%

Supplier: Decon Laboratories Ltd

N0839-25G N0839-5G
TCN0839-25GEA 209 USD InStock
TCN0839-25G TCN0839-5G
4-Nitrochalcone ≥95.0%
4-Nitrochalcone
CAS Number: 1222-98-6
MDL Number: MFCD00007382
Molecular Formula: C15H11NO3
Molecular Weight: 253.26
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 165

Other Product Names / Synonyms: 2-(4-Nitrobenzylidene)acetophenone


Formula: C₁₅H₁₁NO₃
Melting Pt: 165 °C
MDL Number: MFCD00007382
CAS Number: 1222-98-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c1e05b9957231040daee236cf337f05a99d5e73&vid=8c1e05b9957231040daee236cf337f05a99d5e73&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty