P1168-1G
P1168-5G
TCP1168-001GEA
28
USD
InStock
TCP1168-001G
TCP1168-005G
(S)-2-Hydroxy-3-phenylpropanoic acid ≥98.0%
(S)-2-Hydroxy-3-phenylpropanoic acid
CAS Number: 20312-36-1
MDL Number: MFCD00004244
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 124
Specific rotation [a]20/D: -21 deg (C=2, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, (S)-(-)-3-Phenyllactic Acid