Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3-Tris(2-cyanoethoxy)propane ≥97.0%

Supplier: Decon Laboratories Ltd

T0525-500G T0525-25G
TCT0525-500GEA 1588 USD InStock
TCT0525-500G TCT0525-25G
1,2,3-Tris(2-cyanoethoxy)propane ≥97.0%
1,2,3-Tris(2-cyanoethoxy)propane
CAS Number: 2465-93-2
MDL Number: MFCD00019868
Molecular Formula: C12H17N3O3
Molecular Weight: 251.29
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.11

Other Product Names / Synonyms: Glycerol Tris(2-cyanoethyl) Ether


Formula: C₁₂H₁₇N₃O₃
Density: 1.11 (20 °C)
MDL Number: MFCD00019868
CAS Number: 2465-93-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f05a0b7d847444cc4ab0d1c902ad3013c6202787&vid=f05a0b7d847444cc4ab0d1c902ad3013c6202787&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty