Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D(+)-Talose ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

T0869-100MG T0869-500MG
TCT0869-100MGEA 159 USD InStock
TCT0869-100MG TCT0869-500MG
D(+)-Talose ≥98.0% (by HPLC)
D(+)-Talose
CAS Number: 2595-98-4
MDL Number: MFCD00135834
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 134
Specific rotation [a]20/D: 22 deg (C=1, H2O)


Formula: C₆H₁₂O₆
Melting Pt: 134 °C
MDL Number: MFCD00135834
CAS Number: 2595-98-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6707027c35dfd02b898376718abe03910c66807e&vid=6707027c35dfd02b898376718abe03910c66807e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty