046728-03
046728-06
046728-MD
AA46728-03EA
113
USD
InStock
AA46728-03
AA46728-06
AA46728-MD
Polyethylene glycol dimethacrylate, M.W. 3,400
Polyethylene glycol dimethacrylate
Granular solid
CAS No.: 25852-47-5
Molecular Formula: CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Storage Temperature: -30°C to -10°C
Physical Form: Granular solid
Sensitivity: Moisture and Light Sensitive
Appearance: White to off-white
MDL No.: MFCD00081877