Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc formate 98%

Supplier: Origen Biomedical, Inc
Warning

B24210-22 B24210-36
AAB24210-22EA 56.18 USD InStock
AAB24210-22 AAB24210-36
Zinc formate 98%
Zinc formate
Purity: 98% CAS No.: 557-41-5 Molecular Formula: C2H2O4Zn Formula Weight: 155.40 UN No.: 3077 MDL No.: MFCD00050836


Formula: C₂H₂O₄Zn
MW: 155.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050836
CAS Number: 557-41-5
EINECS: 209-176-5
UN: 3077
ADR: 9 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty