Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Cyclopropylethylamine ≥98%, ee 98%, ChiPros®

Supplier: Origen Biomedical, Inc
Danger

H27499-03 H27499-06
AAH27499-03EA 144 USD InStock
AAH27499-03 AAH27499-06
(S)-1-Cyclopropylethylamine ≥98%, ee 98%, ChiPros®
(S)-1-Cyclopropylethylamine
Purity: 98% CAS No.: 195604-39-8 Molecular Formula: C5H11N Formula Weight: 85.15 Flash Point: 3°(37°F) Melting Point: -68° Boiling Point: 95° UN No.: 2733 MDL No.: MFCD08064289


Formula: C₅H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD08064289
CAS Number: 195604-39-8
UN: 2733
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty